Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ BetaBasic™ 4 DASH-HTS HPLC Columns

Catalog No. 71605022150
Click to view available options
Quantity:
1/Pk.
3/Pk.

Designed for high throughput, these short columns are excellent for ballistic gradients, with retention and sensitivity for very fast LC/MS applications

Thermo Scientific HPLC columns are manufactured under strict ISO 9001:2000 procedures, and every batch of silica and bonded phase is thoroughly tested for physical and chromatographic characteristics. Test reports are included with every column shipped.

  • Fully deactivated Type B silica to ensure sharp peaks and excellent reproducibility
  • Give less retention than C18 or C8 phases, and are a good choice for fast analyses including LC/MS of all types of compounds
  • For basic compounds at high pH, which allows for a wide choice of buffer selection in LC/MS applications
  • Stable bonding chemistry and excellent surface coverage gives the columns exceptional low and high pH stability
  • Patented, low dead volume design minimizes band spreading
  • Finger-tight fittings make changing columns quick and easy
  • Available in packs of 3 or packs of 10 for better cost-effectiveness

TRUSTED_SUSTAINABILITY

Specifications

Column Format DASH-HTS Column, 1/Pk.
Diameter (Metric) 2.1 mm
Length (Metric) 20 mm
Particle Size 5 μm
Pore Size 150 Å
Quantity 1/Pk.
USP Type L26
Stationary Phase C4
Packing Material Silica
Carbon Load 6%
Column Type Reversed Phase
Endcapped Yes
Phase Reversed Phase
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.