Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Thermo Scientific™ BetaBasic™ CN SFC Columns
Click to view available options
:
Particle Size: 5μm; 100L x 1.0mm I.D.
Particle Size: 5μm; 100L x 2.1mm I.D.
Particle Size: 5μm; 100L x 4.6mm I.D.
Particle Size: 5μm; 150L x 1.0mm I.D.
Particle Size: 5μm; 150L x 2.1mm I.D.
Particle Size: 5μm; 150L x 4.6mm I.D.
Particle Size: 5μm; 250L x 1.0mm I.D.
Particle Size: 5μm; 250L x 2.1mm I.D.
Particle Size: 5μm; 250L x 4.6mm I.D.
Particle Size: 5μm; 50L x 2.1mm I.D.
Particle Size: 5μm; 50L x 4.6mm I.D.
Description
- Pore size suitable for small molecules, peptides and protein digests
- Superb pH stability

Specifications
Specifications
Diameter (Metric) | 1 mm |
Length (Metric) | 150 mm |
Particle Size | 5 μm |
pH | 2 to 9 |
Pore Size | 150 Å |
Surface Area | 200 m²/g |
USP Type | L10 |
Stationary Phase | Cyano (CN) |
Packing Material | Silica |
Carbon Load | 5% |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction