Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Thermo Scientific™ BioBasic™ Phenyl HPLC Columns
Click to view available options
Length (Metric):
150 mm
Diameter (Metric):
1 mm
4.6 mm
Description
- 300Å pore size for maximum performance with biomolecules
- Lower carbon load for optimal retention of larger peptides and proteins
- Offers different selectivity to alkyl chain chemistires for critical separations
- Excellent for LC/MS separations
Order Info
Other column dimensions are available. Please call Customer Service for more information.

Specifications
Specifications
Diameter (Metric) | 4.6 mm |
Length (Metric) | 150 mm |
Particle Size | 5 μm |
Pore Size | 300 Å |
USP Type | L11 |
Stationary Phase | Phenyl (Ph) |
Packing Material | Silica |
Carbon Load | 3% |
Column Type | Reversed Phase |
Endcapped | Yes |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction