Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ BioBasic™ Phenyl HPLC Columns

Catalog No. p-3616096
Click to view available options
Length (Metric):
150 mm
Diameter (Metric):
1 mm
4.6 mm

Alternative selectivity of peptides and proteins

  • 300Å pore size for maximum performance with biomolecules
  • Lower carbon load for optimal retention of larger peptides and proteins
  • Offers different selectivity to alkyl chain chemistires for critical separations
  • Excellent for LC/MS separations

Order Info

Other column dimensions are available. Please call Customer Service for more information.

TRUSTED_SUSTAINABILITY

Specifications

Diameter (Metric) 1 mm
Length (Metric) 150 mm
Particle Size 5 μm
Pore Size 300 Å
USP Type L11
Stationary Phase Phenyl (Ph)
Packing Material Silica
Carbon Load 3%
Column Type Reversed Phase
Endcapped Yes
Phase Reversed Phase
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.