Learn More
Thermo Scientific™ Maxima SYBR Green/Fluorescein qPCR Master Mix (2X)
Description
Includes
- Maxima SYBR Green/ROX qPCR Master Mix (2x) is supplemented with ROX passive reference dye
- Maxima SYBR Green/Fluorescein qPCR Master Mix (2x) is supplemented with fluorescein passive reference dye
- Maxima SYBR Green qPCR Master Mix, ROX Solution provided (2x), comes with a separate ROX vial
- All Maxima SYBR Green qPCR Master Mixes include Maxima Hot Start Taq DNA Polymerase and dNTPs (also dUTP) in an optimized PCR buffer. Supplied with nuclease-free water
The master mixes include Maxima Hot Start Taq DNA Polymerase and dNTPs in an optimized PCR buffer. Only template and primers need to be added. The SYBR Green I intercalating dye allows for DNA detection and analysis without using sequence-specific probes.
Maxima Hot Start Taq DNA Polymerase in combination with an optimized buffer ensures PCR specificity and sensitivity. dUTP is included in the mix for optional carry-over contamination control using uracil DNA glycosylase (UDG). The use of Maxima SYBR Green qPCR Master Mixes in real-time PCR ensures reproducible, sensitive and specific quantification of genomic, plasmid, viral, and cDNA templates. Maxima SYBR Green qPCR Master Mixes are compatible with most real-time thermal cyclers.
- Specificity - Maxima Hot Start Taq DNA Polymerase and the optimized buffer eliminate non-specific amplification and formation of primer dimers
- Sensitivity - detects low copy number targets
- Wide linear range - accurate quantification across 9 orders of magnitude
- Reproducibility and convenience - ready-to-use 2X master mix
Order Info
Orders placed Monday through Wednesday ship next day; Thursday and Friday orders ship the following Monday.
Specifications
Specifications
| Concentration | 2X |
| Content And Storage | Contains: • 4 x 12.5 mL Maxima SYBR Green/Fluorescein qPCR Master Mix (2X) containing Maxima Hot Start Taq DNA Polymerase, dNTPs (also dUTP) and SYBR Green I in an optimized PCR buffer with fluorescein passive reference dye; sufficient for 4000 x 25 μL reactions. • 2 x 30 mL Nuclease-Free Water Store at -20°C. |
| Detection Method | SYBR |
| Format | Tube |
| GC-Rich PCR Performance | High |
| PCR Method | qPCR |
| Polymerase | Taq DNA Polymerase |
| Reaction Speed | Standard |
| For Use With (Equipment) | BioRad iCycler iQ, BioRad MyiQ, BioRad iQ5 |
| Product Line | Maxima |
| Show More |
For Research Use Only. Not for use in diagnostic procedures.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.