Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TIMP-3 Antibody (277128), mFluor Violet 450 SE, Novus Biologicals™
SDP

Catalog No. NB345623 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB345623 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB345623 Supplier Novus Biologicals Supplier No. FAB9731MFV450
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

TIMP-3 Monoclonal antibody specifically detects TIMP-3 in Human samples. It is validated for Immunoprecipitation, Flow Cytometry, CyTOF-reported

Specifications

Antigen TIMP-3
Applications Immunoprecipitation, Flow Cytometry, CyTOF
Classification Monoclonal
Clone 277128
Conjugate mFluor Violet 450 SE
Dilution Immunoprecipitation, Intracellular Staining by Flow Cytometry, CyTOF-reported
Formulation 50mM Sodium Borate
Gene Alias HSMRK222, K222, K222TA2, metalloproteinase inhibitor 3, MIG-5 protein, Protein MIG-5, pseudoinflammatory), SFD, TIMP metallopeptidase inhibitor 3, TIMP-3, tissue inhibitor of metalloproteinase 3 (Sorsby fundus dystrophy, Tissue inhibitor of metalloproteinases 3
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human TIMP-3, Cys24-Pro211, Accession # P35625
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Angiogenesis, Apoptosis, Cancer, Hypoxia, Metastasis, Vision
Primary or Secondary Primary
Gene ID (Entrez) 7078
Target Species Human
Content And Storage Store at 4C in the dark.
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.