Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TLR3 Antibody, Novus Biologicals™
SDP

Catalog No. NB10056571 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10056571 0.1 mg
NB10056571T 0.025 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB10056571 Supplier Novus Biologicals Supplier No. NB10056571
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody has been used in 16 publications

TLR3 Polyclonal specifically detects TLR3 in Human, Mouse, Porcine samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Dot Blot, Flow (Cell Surface), Flow (Intracellular), Knockdown Validated.

Specifications

Antigen TLR3
Applications Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), Dot Blot, KnockDown
Classification Polyclonal
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1-3 ug/ml, Flow Cytometry 1 ug/10^6 cells, Immunohistochemistry 1:20-1:1000, Immunohistochemistry-Paraffin 1:100. Use reported in scientific literature (Patole et al (2005)), Immunohistochemistry-Frozen 1:20-1:1000. Use reported in scientific literature (Pawar et al (2005)), Dot Blot reported in scientific literature (PMID 27248820), Flow (Cell Surface) reported in scientific literature (Pawar et al (2006)), Flow (Intracellular) 1 ug/1 million cells, KnockDown Validated
Formulation PBS with 0.05% Sodium Azide
Gene Accession No. O15455
Gene Alias CD283, CD283 antigen, toll-like receptor 3
Gene Symbols TLR3
Host Species Rabbit
Immunogen A mix of synthetic peptides corresponding to amino acids 135-150 (SIHKIKSNPFKNQKNL), 828-844 (CRRFKVHHAVQQAIEQN), and 876-891 (CILNWPVQKERINAFH) of mouse TLR3.
Molecular Weight of Antigen 104 kDa
Purification Method Protein G purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Immune System Diseases, Immunology, Innate Immunity, Signal Transduction, Toll Like Receptors
Primary or Secondary Primary
Gene ID (Entrez) 7098
Test Specificity Mouse and Human. Predicted to cross-react with Rat. Porcine data from customer review.
Target Species Human, Mouse, Pig
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.