Learn More
Description
Specifications
Specifications
| Antigen | TLR3 |
| Applications | Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), Dot Blot, KnockDown |
| Classification | Polyclonal |
| Concentration | 1.0 mg/mL |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1-3 ug/ml, Flow Cytometry 1 ug/10^6 cells, Immunohistochemistry 1:20-1:1000, Immunohistochemistry-Paraffin 1:100. Use reported in scientific literature (Patole et al (2005)), Immunohistochemistry-Frozen 1:20-1:1000. Use reported in scientific literature (Pawar et al (2005)), Dot Blot reported in scientific literature (PMID 27248820), Flow (Cell Surface) reported in scientific literature (Pawar et al (2006)), Flow (Intracellular) 1 ug/1 million cells, KnockDown Validated |
| Formulation | PBS with 0.05% Sodium Azide |
| Gene Accession No. | O15455 |
| Gene Alias | CD283, CD283 antigen, toll-like receptor 3 |
| Gene Symbols | TLR3 |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
