Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TLR7 Antibody (4G6) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
$218.50 - $548.00
Specifications
Antigen | TLR7 |
---|---|
Clone | 4G6 |
Concentration | 1 mg/mL |
Dilution | Western Blot 10ug/ml∼, Flow Cytometry 1-2ug/10^6cells, Immunohistochemistry 1:50 - 1:200. Use reported in scientific literature (PMID 31092820), Immunocytochemistry/Immunofluorescence 1:500-1:5000. Use reported in scientific literature (PMID 22610069), Flow (Intracellular) 5 ug/ml, CyTOF-ready, Knockout Validated, KnockDown Validated reported in scientific literature (PMID 31730654) |
Applications | Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, CyTOF, KnockDown |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP227332SS
![]() |
Novus Biologicals
NBP227332SS |
0.025 mg |
Each for $218.50
|
|
|||||
NBP227332
![]() |
Novus Biologicals
NBP227332 |
0.1 mg |
Each for $548.00
|
|
|||||
Description
TLR7 Monoclonal specifically detects TLR7 in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Flow (Intracellular), CyTOF-ready, Knockdown Validated.Specifications
TLR7 | |
1 mg/mL | |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, CyTOF, KnockDown | |
Unconjugated | |
Mouse | |
Cytokine Research, Immunology, Innate Immunity, Toll Like Receptors | |
Q9NYk1 | |
51284 | |
A partial human TLR7 recombinant protein (amino acids 562-839) was used as the immunogen. | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
4G6 | |
Western Blot 10ug/ml∼, Flow Cytometry 1-2ug/10^6cells, Immunohistochemistry 1:50 - 1:200. Use reported in scientific literature (PMID 31092820), Immunocytochemistry/Immunofluorescence 1:500-1:5000. Use reported in scientific literature (PMID 22610069), Flow (Intracellular) 5 ug/ml, CyTOF-ready, Knockout Validated, KnockDown Validated reported in scientific literature (PMID 31730654) | |
Monoclonal | |
Purified | |
RUO | |
Human, Mouse | |
toll-like receptor 7 | |
TLR7 | |
IgG1 κ | |
Protein G purified | |
Human. Mouse reactivity reported in scientific literature (PMID: 24126361). |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title