Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TLR7 Antibody (533707), HRP, Novus Biologicals™
SDP

Catalog No. NB347215 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB347215 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB347215 Supplier Novus Biologicals Supplier No. FAB5875H
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

TLR7 Monoclonal antibody specifically detects TLR7 in Human samples. It is validated for Flow Cytometry, CyTOF

Specifications

Antigen TLR7
Applications Flow Cytometry, CyTOF
Classification Monoclonal
Clone 533707
Conjugate HRP
Dilution Intracellular Staining by Flow Cytometry, CyTOF-ready
Formulation PBS
Gene Alias toll-like receptor 7
Host Species Mouse
Immunogen E. coli-derived recombinant human TLR7, Met360-Leu516, Accession # Q9NYK1
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cytokine Research, Epigenetics, Immune System Diseases, Immunology, Infections (Virus Bacteria and Parasites), Innate Immunity, Signal Transduction, Toll Like Receptors, Virology Bacteria and Parasites
Primary or Secondary Primary
Gene ID (Entrez) 51284
Target Species Human
Content And Storage Store at 4C in the dark.
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.