Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TLR7 Antibody, Novus Biologicals™
SDP

Catalog No. p-5094746 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10056588T 0.025 mg
NB10056588 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB10056588T Supplier Novus Biologicals Supplier No. NB10056588SS
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody has been used in 13 publications

TLR7 Polyclonal specifically detects TLR7 in Human, Mouse samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Flow (Cell Surface), Flow (Intracellular).

Specifications

Antigen TLR7
Applications Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen)
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Flow Cytometry, Immunohistochemistry 1:10 - 1:500, Immunohistochemistry-Paraffin 1:10 - 1:50, Immunohistochemistry-Frozen 1:20. Use 1:20, Flow (Cell Surface) reported in scientific literature (Wong et al (2007)), Flow (Intracellular) 0.5 - 2 ug/1x10^6 cells
Formulation PBS with 0.05% Sodium Azide
Gene Accession No. Q9NYK1
Gene Alias toll-like receptor 7
Gene Symbols TLR7
Host Species Rabbit
Immunogen This antibody was developed against KLH-conjugated synthetic peptide corresponding to a portion of amino acids 684-701 of human TLR7 (NP_057646).
Purification Method Immunogen affinity purified
Quantity 0.025 mg
Regulatory Status RUO
Research Discipline Immunology, Innate Immunity, Toll Like Receptors
Primary or Secondary Primary
Gene ID (Entrez) 51284
Test Specificity Reacts with Human and Mouse. 72% sequence identity with rat protein.
Target Species Human, Mouse
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.