Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TLR7 Antibody, Novus Biologicals™
SDP

Catalog No. p-5094750 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mL
0.1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10056682T 0.025 mL
NB10056682 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB10056682T Supplier Novus Biologicals Supplier No. NB10056682SS
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody has been used in 4 publications

TLR7 Polyclonal specifically detects TLR7 in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.

Specifications

Antigen TLR7
Applications Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1.0 mg/ml
Conjugate Unconjugated
Dilution Western Blot 1:500-1:1000, Flow Cytometry, Immunohistochemistry reported in scientific literature (PMID 28028829), Immunocytochemistry/Immunofluorescence reported in scientific literature (PMID 24886842), Immunohistochemistry-Paraffin 1:100
Formulation PBS with 0.05% Sodium Azide
Gene Accession No. NP_057646
Gene Alias toll-like receptor 7
Gene Symbols TLR7
Host Species Rabbit
Immunogen This antibody was developed against KLH-conjugated synthetic peptide corresponding to amino acids 465-483 of human TLR7.
Purification Method Protein G purified
Quantity 0.025 mL
Regulatory Status RUO
Research Discipline Immunology, Innate Immunity
Primary or Secondary Primary
Gene ID (Entrez) 51284
Target Species Human, Mouse
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.