Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Toxoplasma Gondii P30 Antibody (P30/3) - BSA Free, Novus Biologicals™
SDP

Catalog No. NB10066486 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10066486 0.1 mg
NB017776 0.025 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB10066486 Supplier Novus Biologicals Supplier No. NB10066486

Mouse Monoclonal Antibody has been used in 3 publications

Toxoplasma Gondii P30 Monoclonal specifically detects Toxoplasma Gondii P30 in Protozoa, T. gondii samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen Toxoplasma Gondii P30
Applications Western Blot, Flow Cytometry, ELISA, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone P30/3
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:100-1:2000, Flow Cytometry 1:10-1:1000, ELISA 1:100-1:2000, Immunocytochemistry/Immunofluorescence
Gene Alias p30, SAG1, SAG1 protein
Host Species Mouse
Immunogen Purified T. gondii P30 antigen
Purification Method Protein G purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity Specific for the Toxoplasma gondii P30 membrane protein.
Target Species Protozoa, T. gondii
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2a
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.