Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TPPP3 Antibody, mFluor Violet 610 SE, Novus Biologicals™
SDP

Catalog No. NB329097 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB329097 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB329097 Supplier Novus Biologicals Supplier No. NBP300282MFV610
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

TPPP3 Polyclonal antibody specifically detects TPPP3 in Human samples. It is validated for ELISA

Specifications

Antigen TPPP3
Applications ELISA
Classification Polyclonal
Conjugate mFluor Violet 610 SE
Dilution ELISA
Formulation 50mM Sodium Borate
Gene Alias brain specific protein, CGI-38, p20, p25gamma, TPPP/p20, tubulin polymerization-promoting protein family member 3
Host Species Rabbit
Immunogen Produced in rabbits immunized with purified, recombinant Human TPPP3 (Uniprot#: Q9BW30; Met1-Lys176)
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Biology, Cell Cycle and Replication, Cytoskeleton Markers, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 51673
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.