Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TPR Antibody, Novus Biologicals™
SDP

Catalog No. NB1002864 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
Catalog No. Quantity
NB1002864 0.1 mL
1 options

Catalog No. NB1002864

Supplier: Novus Biologicals NB1002864

Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

TPR Polyclonal specifically detects TPR in Human samples. It is validated for Western Blot, Immunoprecipitation.

Specifications

Antigen TPR
Applications Western Blot, Immunoprecipitation
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:2000-1:10000, Immunoprecipitation 2-5 ug/mg lysate
Formulation TBS and 0.1% BSA with 0.09% Sodium Azide
Gene Accession No. P12270
Gene Alias nuclear pore complex-associated protein TPR, nucleoprotein TPR, translocated promoter region (to activated MET oncogene), tumor potentiating region
Gene Symbols TPR
Host Species Rabbit
Immunogen The immunogen recognized by this antibody maps to a region between residue 600 and 650 of human Translocated Promoter Region Protein using the numbering given in entry NP_003283.1 (GeneID 7175).
Molecular Weight of Antigen 267 kDa
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7175
Target Species Human
Content And Storage Store at 4C. Do not freeze.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.