Learn More
Description
Specifications
Specifications
| Antigen | TRAF-2 |
| Applications | Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1:1000-1:2000, Flow Cytometry, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunoprecipitation 1:50-1:200, Immunohistochemistry-Paraffin 1:1000-1:5000 |
| Formulation | Whole antisera with 0.02% Sodium Azide |
| Gene Alias | E3 ubiquitin-protein ligase TRAF2, EC 6.3.2.-, MGC:45012, TNF receptor-associated factor 2, TRAP3TRAP, tumor necrosis factor type 2 receptor associated protein 3, Tumor necrosis factor type 2 receptor-associated protein 3 |
| Gene Symbols | TRAF2 |
| Host Species | Rabbit |
| Immunogen | A synthetic peptide corresponding to amino acids 6-31 (VTPPGSLELLQPGFSKTLLGTKLEAK) of human TRAF2 was used as immunogen, GenBank no. NP_066961.2. This peptide sequence is 92% conserved in mouse and rat TRAF2. The peptide immunogen sequence is 100% conserved in multiple human TRAF2 GenBank accession numbers. These TRAF2 accession numbers are of varying amino acid (aa) lengths and include: 1) NP_066961.2 = 501 aa. 2) CAI12705.1 =121 aa. 3) CAI12704.1 = 422 aa. 4) CAI15104.1 = 193 aa. 5) BAD96681.1 = 205 aa. |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
