Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TRF-2 Antibody - BSA Free, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 86 publications
$279.00 - $594.00
Specifications
Antigen | TRF-2 |
---|---|
Concentration | 1.0 mg/mL |
Dilution | Western Blot 1:2000 - 1:5000, Simple Western 1:25, Flow Cytometry 1-5 ug/ml, ELISA reported in scientific literature (PMID 31575660), Immunohistochemistry 1:200, Immunocytochemistry/Immunofluorescence 1:50 - 1:200, Immunoprecipitation 1:10 - 1:500. Use reported in scientific literature, Immunohistochemistry-Paraffin 1:200, Dot Blot reported in scientific literature (PMID 31026066), Chromatin Immunoprecipitation (ChIP) 1:10-1:500, KnockDown Validated reported in scientific literature (PMID 31026066) |
Applications | Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin), Dot Blot, KnockDown |
Classification | Polyclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB11057130T
![]() |
Novus Biologicals
NB11057130SS |
0.025 mL |
Each for $279.00
|
|
|||||
NB11057130
![]() |
Novus Biologicals
NB11057130 |
0.1 mL |
Each for $594.00
|
|
|||||
Description
TRF-2 Polyclonal specifically detects TRF-2 in Human, Mouse, Rat, Chinese Hamster, Primate samples. It is validated for Western Blot, Simple Western, Chromatin Immunoprecipitation, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Dot Blot, Knockdown Validated.Specifications
TRF-2 | |
Western Blot 1:2000 - 1:5000, Simple Western 1:25, Flow Cytometry 1-5 ug/ml, ELISA reported in scientific literature (PMID 31575660), Immunohistochemistry 1:200, Immunocytochemistry/Immunofluorescence 1:50 - 1:200, Immunoprecipitation 1:10 - 1:500. Use reported in scientific literature, Immunohistochemistry-Paraffin 1:200, Dot Blot reported in scientific literature (PMID 31026066), Chromatin Immunoprecipitation (ChIP) 1:10-1:500, KnockDown Validated reported in scientific literature (PMID 31026066) | |
Polyclonal | |
Rabbit | |
Cancer, Cell Cycle and Replication, Cellular Markers, Chromatin Research, Transcription Factors and Regulators | |
PBS with 0.02% Sodium Azide | |
Telomeric DNA-binding protein, telomeric repeat binding factor 2, Telomeric repeat binding protein 2, telomeric repeat-binding factor 2, TERF 2, TERF2, TRBF 2, TRBF2, TRF 2, TRF2, TTAGGG repeat binding factor 2, TTAGGG repeat-binding factor 2 | |
TERF2 | |
IgG | |
Affinity Purified | |
59.6 kDa |
1.0 mg/mL | |
Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin), Dot Blot, KnockDown | |
Unconjugated | |
RUO | |
Human, Mouse, Rat, Chinese Hamster, Primate | |
Q15554 | |
7014 | |
This TRF-2 Antibody was developed against Baculovirus purified TRF2 protein. | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title