Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Triosephosphate isomerase Antibody, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody

$158.00 - $499.50

Specifications

Antigen Triosephosphate isomerase
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Host Species Rabbit
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP1528820
SDP
View Documents
Novus Biologicals
NBP15288120UL
20 μL
Each for $158.00
Only null left
Add to Cart
 
NBP152881
SDP
View Documents
Novus Biologicals
NBP152881
100 μL
Each for $499.50
Only null left
Add to Cart
 
Description

Description

Triosephosphate isomerase Polyclonal specifically detects Triosephosphate isomerase in Human samples. It is validated for Western Blot.
Specifications

Specifications

Triosephosphate isomerase
Polyclonal
Rabbit
Stem Cell Markers
EC 5.3.1.1, MGC88108, TIM, TPI, triosephosphate isomerase, Triose-phosphate isomerase, triosephosphate isomerase 1
TPI1
IgG
27 kDa
Western Blot
Unconjugated
RUO
P60174
7167
Synthetic peptides corresponding to TPI1(triosephosphate isomerase 1) The peptide sequence was selected from the N terminal of TPI1. Peptide sequence MAPSRKFFVGGNWKMNGRKQSLGELIGTLNAAKVPADTEVVCAPPTAYID.
Primary
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.