Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tropomyosin-1, Rat anti-Mouse, Human, Clone: 7F8B8, Millipore Sigma™
Rat Monoclonal Antibody
$192.34 - $482.80
Specifications
Antigen | Tropomyosin-1 |
---|---|
Clone | 7F8B8 |
Dilution | Immunocytochemistry Analysis: A representative lot detected Tropomyosin-1 in Immunocytochemistry applications (Brayford, S., et. al. (2016). Curr Biol. 26(10):1312-8). |
Applications | Immunocytochemistry, Western Blot |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MABT136525
|
MilliporeSigma
MABT136525UL |
25 μL |
Each for $192.34
|
|
|||||
MABT1365100
|
MilliporeSigma
MABT1365100UL |
100 μL |
Each for $482.80
|
|
|||||
Description
Anti-Tropomyosin-1, clone 7F8B8, Cat. No. MABT1365, is a rat monoclonal antibody that detects Tropomyosin-1 and has been tested for use in Immunocytochemistry and Western Blotting.
Tropomyosin alpha-1 chain (UniProt: P09493; also known as Alpha-tropomyosin, Tropomyosin-1) is encoded by the TPM1 (also known as C15orf13, TMSA) gene (Gene ID: 7168) in human. Tropomyosin-1 is a cytoskeletal protein that associates with F-actin stress fibers. It binds to actin filaments in muscle and non-muscle cells and plays a central role, in association with the troponin complex, in the calcium dependent regulation of vertebrate striated muscle contraction. It stabilizes actin filaments and regulates the interaction of actin filament with other actin-binding proteins. In non-muscle cells is implicated in stabilizing cytoskeleton actin filaments. Tropomyosin-1 is a homodimeric protein, but can form heterodimer of an alpha (TPM1, TPM3, or TPM4) and a beta (TPM2) chain. Mutations in TPM1 gene have been linked to cardiomyopathy characterized by ventricular hypertrophy and ventricular dilation and impaired systolic function that results in congestive heart failure and arrhythmia. Ten different isoforms of Tropomyosin-1 have been described that are produced by alternative splicing.Specifications
Tropomyosin-1 | |
Immunocytochemistry Analysis: A representative lot detected Tropomyosin-1 in Immunocytochemistry applications (Brayford, S., et. al. (2016). Curr Biol. 26(10):1312-8). | |
Monoclonal | |
Purified | |
RUO | |
Mouse, Human | |
Tropomyosin alpha-1 chain;Alpha-tropomyosin;Tropomyosin-1 | |
TPM1;C15orf13;TMSA | |
IgG2a κ | |
Protein G purified | |
Clone 7F8B8 detects human and murine Tropomyosin-1. It targets an epitope with in the N-terminal region. |
7F8B8 | |
Immunocytochemistry, Western Blot | |
Unconjugated | |
Rat | |
Cell Structure | |
Purified rat monoclonal antibody IgG2a in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide. | |
NP_000357.3 | |
KLH-conjugated linear peptides corresponding to 24 amino acids from the N-terminal region of isoform 5 and 4 amino acids from the N-terminal region of isoform 2 of human tropomyosin-1. | |
Primary | |
Stable for 1 year at 2-8°C from date of receipt. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title