Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tropomyosin alpha-1 Mouse anti-Rat, Mouse, Clone: 3F5E4, MilliporeSigma™
Mouse Monoclonal Antibody
$191.34 - $481.79
Specifications
| Antigen | Tropomyosin alpha-1 |
|---|---|
| Clone | 3F5E4 |
| Concentration | 0.5 mg/mL |
| Applications | Western Blot |
| Classification | Monoclonal |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|
MABT133825
|
MilliporeSigma
MABT133825UL |
25 μL |
Each for $191.34
|
|
|||||
|
MABT1338100
|
MilliporeSigma
MABT1338100UL |
100 μL |
Each for $481.79
|
|
|||||
Description
Tropomyosin alpha-1 Monoclonal antibody specifically detects Tropomyosin alpha-1 in Rat, Mouse samples. It is validated for Western Blotting.Specifications
| Tropomyosin alpha-1 | |
| 0.5 mg/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Purified mouse monoclonal antibody IgG1 in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide. | |
| Primary | |
| Stable for 1 year at 2-8°C from date of receipt. |
| 3F5E4 | |
| Western Blot | |
| Unconjugated | |
| Mouse | |
| Rat, Mouse | |
| KLH-conjugated linear peptide corresponding to 24 amino acids from the N-terminal region of rat Tropomyosin alpha-1 chain, isoform 2. | |
| Protein G purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title