Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TRPV6 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$158.00 - $489.50
Specifications
Antigen | TRPV6 |
---|---|
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Regulatory Status | RUO |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP17413820
![]() |
Novus Biologicals
NBP17413820UL |
20 μL |
Each for $158.00
|
|
|||||
NBP174138
![]() |
Novus Biologicals
NBP174138 |
100 μL |
Each for $489.50
|
|
|||||
Description
TRPV6 Polyclonal specifically detects TRPV6 in Human, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
TRPV6 | |
Unconjugated | |
RUO | |
ABP/ZF, calcium channel CaT1, Calcium transport protein 1, CaT1, CaT-L, CATL, CaT-like, ECaC2, ECAC2caT-L, epithelial apical membrane calcium transporter/channel CaT1, Epithelial calcium channel 2Alu-binding protein with zinc finger domain, HSA277909, LP6728, transient receptor potential cation channel subfamily V member 6, transient receptor potential cation channel, subfamily V, member 6, TrpV6, ZFAB | |
TRPV6 | |
IgG | |
83 kDa |
Polyclonal | |
Rabbit | |
Q9R186 | |
55503 | |
Synthetic peptides corresponding to the middle region of Trpv6. Immunizing peptide sequence TEIDSSGDDQSLLELIVTTKKREARQILDQTPVKELVSLKWKRYGRPYFC. | |
Primary |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title