Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Applied Biosystems™ TrueMark™ PGx Comprehensive Plasmid Controls

Catalog No. a65342
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 Kit, 15 Tubes total
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A65342 1 Kit, 15 Tubes total
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A65342 Supplier Applied Biosystems™ Supplier No. A65342
Only null left
Add to Cart
Add to Cart

TrueMark PGx Comprehensive Plasmid Controls contains three linearized multi-target plasmid pools with human genome target sequences commonly used for pharmacogenomic research testing.

TrueMark PGx Comprehensive Plasmid Controls contains three linearized multi-target plasmid pool controls with human genome target sequences commonly used for pharmacogenomics research testing.

These controls were designed for use with TaqMan real-time qPCR assays to verify the presence of homozygous and heterozygous genotyping, copy number variation (CNV), and CNV reference assays (TERT and RNase P). This control kit provides coverage of essential, commonly studied markers within CYP2D6, CYP2C19, CYP2C9, DPYD, and other important DME and CNV research genes.

The three controls are supplied at a concentration of ~1 x 105 copies/μL in 5 tubes of 50 μL each. They are quantified using digital PCR for accuracy and can be used as stand-alone samples to assess assay performance or to assist with assay troubleshooting. They may also serve as a CNV positive run control/calibrator.

Compatible OpenArray and TaqMan array plates/panels

  • TaqMan OpenArray PGx Express 60 Panel, Cat. No. 4488847
  • TaqMan PGx Express Panel (64), 384-well Plate, Cat. No. A37610
  • TaqMan OpenArray PGx Express 120 Panel (v3)
  • TrueMark PGx SNP + CNV, 64 Panel, 384-well plate, Cat. No. A40003215
  • TrueMark PGx SNP + CNV, 64 Panel, OpenArray
  • TrueMark PGx SNP + CNV, 128 Panel, OpenArray

If your assay is not supported with this product, please consider the TaqMan Custom DNA Controls offered by GeneArt. This product is manufactured in an ISO 9001 compliant facility.

TRUSTED_SUSTAINABILITY

Specifications

Concentration 1 x 105 Copies/μL
Detection Method TaqMan
Form Frozen
PCR Method qPCR, Digital PCR
Technique Real-time qPCR
Quantity 1 Kit, 15 Tubes total
Shipping Condition Dry Ice
For Use With (Application) Genotyping, Copy Number Variation Analysis, Real Time PCR (qPCR), Digital PCR (dPCR)
No. of Reactions 3 control pools (5 aliquiots x 50 μL each pool)
Volume 50 μL

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.