Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TSKgel™ G2000PW HPLC Column, 12 μm Particle Size, Sold by MilliporeSigma™ Supelco™

Catalog No. p-7213958 Shop All MilliporeSigma Supelco Products
Click to view available options
Length (Metric):
30 cm
60 cm

Use the TSKgel™ G2000PW HPLC column for analysis of water-soluble polymers with molecular weights up to 2,000 Da.

Order Info

Shipped in water.

TRUSTED_SUSTAINABILITY

Specifications

Diameter (Metric) 7.5 mm
Flow Rate 1.2 mL/min.
Length (Metric) 30 cm
Max. Pressure 20 bar
Particle Size 12 μm
pH 2.0 - 12
Pore Size 125 Å
Solvents ≤20% organic solvent
Temperature 80°C
USP Type L25, L38, L39
Stationary Phase Hydroxylated Methacrylate
Product Line TSKgel
Matrix Polymer Particle Platform
Phase Size Exclusion
Show More Show Less

TSKgel is a registered trademark of Tosoh Corporation

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.