Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TSKgel™ SP-5PW HPLC Column, Sold by MilliporeSigma™ Supelco™

Catalog No. p-7089812 Shop All MilliporeSigma Supelco Products
Click to view available options
Diameter (Metric):
2 mm
5 mm
8 mm
20 mm
Material:
Glass
Stainless Steel Hardware

For the separation and analysis of proteins, peptides, and other biologically active molecules.

  • Packed with porous hydrophilic polymer beads that are surface modified with a strong cation exchanger
  • For the separation and analysis of proteins, peptides, and other biologically active molecules
  • Columns have the same backbone chemistry and selectivity as the bulk process scale resins Toyopearl SP-650 & TSKgel SP-5PW (20) and (30)
  • Salt Concentration: ≤1 M
TRUSTED_SUSTAINABILITY

Specifications

Diameter (Metric) 5 mm
Flow Rate 1 mL/min
Ion Exchange Group Na+
Length (Metric) 5 cm
Max. Pressure 150 bar
Particle Size 10 μm
pH 2 to 12
Solvents ≤ 20% Organic Solvent
Temperature 10°C to 45°C
Stationary Phase Sulfonic Acid
Product Line TSKgel
Matrix Sulfonic Acid Phase/Hydroxylated Methacrylic Polymer
Material Glass
Technique Cation Exchange
Show More Show Less

TSKgel is a registered trademark of Tosoh Corporation

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.