Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TSKgel™ Super-ODS HPLC Column, 2.3 μm Particle Size, Sold by MilliporeSigma™ Supelco™

Catalog No. 111021929 Shop All MilliporeSigma Supelco Products
Click to view available options
Length (Metric):
5 cm
10 cm
Diameter (Metric):
1 mm
2 mm
4.6 mm

TSKgel™ Super-ODS Columns are packed with monodispersed, 2 μm (nominal, mean particle size is 2.3 micron) spherical silica particles covalently bonded with phenyl groups.

  • The small particle size used in Super Series and TSKgel ODS-140HTP Columns provides the highest efficiency of all TSKgel reversed phase columns
  • Monodispersity of the particles reduces column back pressure to values more typical of larger particles, allowing the use of higher flow rates compared to other 2 μm packings
  • TSKgel Super-ODS Columns are more hydrophobic than TSKgel Super-Octyl and TSKgel Super-Phenyl Columns
  • An excellent choice for peptides, proteins, amino acids, tryptic digests, nucleotides, pharmaceutical molecules, and food/beverage samples
  • Shipped in 70% Methanol, 30% Water
TRUSTED_SUSTAINABILITY

Specifications

Diameter (Metric) 4.6 mm
Flow Rate 4 mL/min
Length (Metric) 5 cm
Max. Pressure 300 bar
Particle Size 2.3 μm
pH 2 to 7.5
Pore Size 140 Å
Solvents 0-100% Organic Solvent
Surface Area 100 m2/g
Temperature 10°C to 50°C
USP Type L1
Stationary Phase C18 (octadecyl)
Product Line TSKgel
Shipping Condition Shipped in 70% Methanol, 30% Water
Matrix C18 (octadecyl) Phase/Silica Particle Platform
Carbon Load 8%
Endcapped Yes
Phase Reversed Phase
Show More Show Less

TSKgel is a registered trademark of Tosoh Corporation

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.