Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TWIST Monoclonal Antibody, Active Motif™
SDP

Mouse Monoclonal Antibody

$132.55 - $397.65

Specifications

Antigen TWIST
Concentration 1 mg/mL
Applications Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Western Blot
Classification Monoclonal
Conjugate Unconjugated
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
50-199-3140
SDP
Encompass
View Documents
Active Motif
61098
10 μg
Each for $132.55
Only null left
Add to Cart
 
50-199-3139
SDP
Encompass
View Documents
Active Motif
61097
100 μg
Each for $397.65
Only null left
Add to Cart
 
Description

Description

TWIST1 acts as a transcriptional regulator. It inhibits myogensis by sequestering E proteins, inhibiting trans-activation by MEF2, and inhibiting DNA-binding by MYOD1 through physical interaction. TWIST1 also represses proinflammatory cytokines such as TNFA and IL1B. TWIST also regulates cranial suture patterning and fusion and gene expression differentially depending on dimer composition. Mutations in the gene are implicated in Saethre-Chotzen syndrome, a disease characterized by coronal synostosis, brachycephaly, low front hairline, facial asymmetry, hypertelorism, broad halluces, and clinodactyly.
Specifications

Specifications

TWIST
Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Western Blot
Unconjugated
Mouse
Human, Mouse
P26687, Q15672
22160, 7291
This TWIST antibody was raised against a recombinant protein corresponding to full-length mouse TWIST.
Primary
-20°C, Avoid Freeze/Thaw Cycles
TWIST1
1 mg/mL
Monoclonal
Liquid
RUO
70mM tris with 0.07mM EDTA, 105mM NaCl, 30% glycerol, 31mM glycine and 0.035% sodium azide; pH 8
AA960487; ACS3; B-HLH DNA binding protein; BHLHA38; BOS_4079; BPES2; BPES3; charlie chaplin; Class A basic helix-loop-helix protein 38; CRS; CRS1; CSO; H-twist; M-twist; p ACS3; Pde; pdt; Pluridigite; polydactyly EMS; SCS; Ska; Skam10Jus; Ska10; Twist; twist basic helix-loop-helix transcription factor 1; twist family bHLH transcription factor 1; twist gene homolog 1; twist homolog 1; twist homolog 1 (Drosophila); TWIST homolog of drosophila; TWIST1; Twist-related protein 1
TWIST1
IgG1
Protein G
Antibody
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.