Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ U937 Whole Cell Lysate
SDP

Catalog No. NBP20396001 Shop All Novus Biologicals Products
Click to view available options
Quantity:
0.1 mg

U937 (Human Histiocytic Lymphoma) whole blood lymph cell lysate 2mg/mL supplied in SDS sample buffer containing 5% Beta-mercaptoethanol. Growth media: DMEM and 10% NCS (newborn Calf serum). Lysates are guaranteed for 6 months from date of receipt.

The U937 Whole Cell Lysate is derived from human. It has been validated for the following applications: Western Blot.

Specifications

For Use With (Application) Western Blot
Tissue U937
Quantity 0.1 mg
Storage Buffer Supplied in SDS sample buffer containing 5% Beta-mercaptoethanol.
Test Specificity U937 (Human Histiocytic Lymphoma)
Host Species Human
Concentration 2 mg/mL
Content And Storage Store at −80°C. Avoid freeze-thaw cycles.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.