Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Ubiquitin Antibody (Ubi-1), Novus Biologicals™

Mouse Monoclonal Antibody has been used in 32 publications
$267.71 - $618.28
Specifications
| Antigen | Ubiquitin |
|---|---|
| Clone | Ubi-1 |
| Concentration | 1 mg/ml |
| Dilution | Western Blot 1:1000-1:2000, Immunohistochemistry 1:2000, Immunocytochemistry/Immunofluorescence 1:2000, Immunohistochemistry-Paraffin 1:2000, Immunohistochemistry-Frozen 1:2000, Immunoblotting |
| Applications | Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), Immunoblot |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB300130SS
![]() |
Novus Biologicals
NB300130SS |
0.025 mL |
Each for $267.71
|
|
|||||
NB300130
![]() |
Novus Biologicals
NB300130 |
0.1 mL |
Each for $618.28
|
|
|||||
Description
Ubiquitin Monoclonal specifically detects Ubiquitin in Human, Mouse, Rat, Porcine, Bovine, C. elegans, Chicken, Drosophila, Equine, Plant, Monkey, Zebrafish samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Immunoblotting.Specifications
| Ubiquitin | |
| 1 mg/ml | |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), Immunoblot | |
| Unconjugated | |
| Mouse | |
| Human, Mouse, Rat, Pig, Bovine, C. elegans, Chicken, Drosophila, Equine, Plant, Monkey, Zebrafish | |
| P0CG48, P0CG47 | |
| 6233 | |
| Purified bovine erythrocyte ubiquitin cross-linked with glutaraldehyde to KLH | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| 8.5 kDa |
| Ubi-1 | |
| Western Blot 1:1000-1:2000, Immunohistochemistry 1:2000, Immunocytochemistry/Immunofluorescence 1:2000, Immunohistochemistry-Paraffin 1:2000, Immunohistochemistry-Frozen 1:2000, Immunoblotting | |
| Monoclonal | |
| Ascites | |
| RUO | |
| 50% PBS, 50% glycerol with 5mM Sodium Azide | |
| RPS27A, UBA52, UBB ubiquitin B, UBC | |
| RPS27A | |
| IgG1 | |
| Affinity purified | |
| Recognizes polyubiquitin chains much more strongly than monoubiquitinated molecules or free ubiquitin. Specifically recognizes ubiquitinated cytoplasmic inclusion bodies. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title