Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
UBXN2A Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$206.00 - $487.50
Specifications
Antigen | UBXN2A |
---|---|
Dilution | Western Blot 1.0 ug/ml |
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP17416220
![]() |
Novus Biologicals
NBP17416220UL |
20 μL |
Each for $206.00
|
|
|||||
NBP174162
![]() |
Novus Biologicals
NBP174162 |
100 μL |
Each for $487.50
|
|
|||||
Description
UBXN2A Polyclonal specifically detects UBXN2A in Rat samples. It is validated for Western Blot.Specifications
UBXN2A | |
Western Blot | |
Unconjugated | |
RUO | |
MGC138202, UBX domain containing 4, UBX domain protein 2A, UBX domain-containing protein 2A, UBX domain-containing protein 4, UBXD4 | |
UBXN2A | |
IgG | |
29 kDa |
Western Blot 1.0 ug/ml | |
Polyclonal | |
Rabbit | |
D3ZID8 | |
165324 | |
Synthetic peptides corresponding to the middle region of Ubxn2a. Immunizing peptide sequence VCMSTKPVFQPFSGQGHRLGSATPRIVSKAKSIEVDNKSTLSAVSLNNLE. | |
Primary |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title