Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ULBP-4/RAET1E Antibody (709116), mFluor Violet 610 SE, Novus Biologicals™
SDP

Catalog No. NB347050 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB347050 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB347050 Supplier Novus Biologicals Supplier No. FAB6285MFV610
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

ULBP-4/RAET1E Monoclonal antibody specifically detects ULBP-4/RAET1E in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, CyTOF

Specifications

Antigen ULBP-4/RAET1E
Applications Western Blot, Flow Cytometry, Immunohistochemistry, CyTOF
Classification Monoclonal
Clone 709116
Conjugate mFluor Violet 610 SE
Dilution Western Blot, Flow Cytometry, Immunohistochemistry, CyTOF-ready
Formulation 50mM Sodium Borate
Gene Alias bA350J20.7, LETALRAE-1-like transcript 4, Lymphocyte effector toxicity activation ligand, MGC125309, N2DL4, N2DL-4, NKG2D ligand 4, NKG2DL4, RAET1E2, retinoic acid early transcript 1ERL-4, ULBP4MGC125308
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human ULBP-4/RAET1E, Gly30-Asp225, Accession # Q8TD07
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Gene ID (Entrez) 135250
Target Species Human
Content And Storage Store at 4C in the dark.
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.