Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

UNC93B Antibody, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody has been used in 2 publications

$206.50 - $499.50

Specifications

Antigen UNC93B
Concentration 1.0 mg/mL
Dilution Western Blot 3-5 ug/ml, Immunocytochemistry/Immunofluorescence reported in scientific literature (PMID 26067373)
Applications Western Blot, Immunocytochemistry, Immunofluorescence
Classification Polyclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP224743SS
SDP
View Documents
Novus Biologicals
NBP224743SS
0.025 mg
Each for $206.50
Only null left
Add to Cart
 
NBP224743
SDP
View Documents
Novus Biologicals
NBP224743
0.1 mg
Each for $499.50
Only null left
Add to Cart
 
Description

Description

UNC93B Polyclonal specifically detects UNC93B in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.
Specifications

Specifications

UNC93B
Western Blot 3-5 ug/ml, Immunocytochemistry/Immunofluorescence reported in scientific literature (PMID 26067373)
Polyclonal
Purified
RUO
0.1 ml PBS with 0.05% Sodium Azide
81622
A portion of amino acid 500-550 of mouse UNC93B protein was used as the immunogen.
Primary
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
1.0 mg/mL
Western Blot, Immunocytochemistry, Immunofluorescence
Unconjugated
Rabbit
Human, Mouse, Rat
hUNC93B1, MGC126617, unc93 (C. elegans) homolog B1, unc93 homolog B, unc93 homolog B1, unc-93 homolog B1 (C. elegans), unc-93 related protein, UNC93B, Unc-93B1, UNC93protein unc-93 homolog B1
UNC93B1
IgG
Protein G purified
The amino acid sequence used as immunogen is 100% homologous in mouse, 77% homologous in opossum and 72% homologous in xenopus.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.