Learn More
Honeywell™ Uvex™ Millennia™ Protective Safety Glasses
Futuristic wraparound styling with exceptional coverage and protection
Specifications
Certifications/Compliance | Meet ANSI Z87.1-2003 and CSA Z94.3 standards as well as military V0 ballistic test for impact. |
---|---|
Lens Material | Polycarbonate |
Catalog Number | Mfr. No. | Frame Color | Lens Coating | Lens Tint | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Frame Color | Lens Coating | Lens Tint | Price | Quantity & Availability | |||
19-039-229
|
Honeywell™
11150362 |
Gray | Scratch-resistant | Silver Mirror | N/A | N/A | N/A | ||
19-087-375B
|
Honeywell™
11150356 |
Black | Impact-resistant, Scratch-resistant | Gray | N/A | N/A | N/A | ||
19-039-212
|
Honeywell™
11150354 |
Black | Scratch-resistant | Silver Mirror | N/A | N/A | N/A | ||
19-039-213
|
Honeywell™
11150370 |
Blue | Impact-resistant, Scratch-resistant | Clear | N/A | N/A | N/A | ||
19-087-375A
|
Honeywell™
11150355 |
Black | Impact-resistant | Clear | N/A | N/A | N/A | ||
Description
- Lightweight nylon frame with polycarbonate lens
- Superior side and brow protection
- High impact dual 9-base wraparound unilens
- Comfortable universal nose bridge
Order Info
Lenses with Fog-Ban™ coating available for clear vision in humid ambient conditions.
Compliance
Meet ANSI™ Z87.1-2010 and CSA Z94.3 standards as well as military V0 ballistic test for impact.
Specifications
Meet ANSI Z87.1-2003 and CSA Z94.3 standards as well as military V0 ballistic test for impact. | |
Polycarbonate |