Learn More
Honeywell™ Uvex™ Strategy™ Safety Goggles
Economical over-the-glass (OTG) goggle that delivers both premium comfort and maximum protection
Specifications
Lens Coating Anti-fog
Lens Type Uvextra
Lens Tint Clear
View More Specs
Products 5
Catalog Number Mfr. No. Frame Color Includes Ventilation Details Price Quantity & Availability  
Catalog Number Mfr. No. Frame Color Includes Ventilation Details Price Quantity & Availability  
19-167-203
View Documents
Honeywell™
S3810
Gray Neoprene band Indirect vent Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please to view product availability in your area.
View Alternate Product
N/A
19-167-201
View Documents
Honeywell™
S3800
Gray Fabric band Direct vent Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please to view product availability in your area.
View Alternate Product
N/A
19-167-202
View Documents
Honeywell™
S3805
Gray Neoprene band Closed vent Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please to view product availability in your area.
View Alternate Product
N/A
19-167-204
View Documents
Honeywell™
S3815
Gray Fabric band Indirect vent with foam Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please to view product availability in your area.
View Alternate Product
N/A
19-167-205
View Documents
Honeywell™
S3820
Orange Neoprene band Indirect vent Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please to view product availability in your area.
View Alternate Product
N/A
Description
Description
Provide protection against impact, chemical splash, dust, sand and debris — even in extreme environments. Available in four models for a variety of work applications: direct vent, closed vent, indirect vent and indirect vent with foam lining for fine particle protection.
  • Ideal for most indoor work applications
  • Rugged design that withstands the harshest environments
  • Lens tint: clear
  • Lenses offer 99.9% ultra-violet protection
  • Headband available in fabric or neoprene for chemical applications
  • Uvextra™ anti-fog coating minimizes fogging
  • Lightweight, TPE body provides exceptional comfort and a snug fit for maximum protection
  • Fits comfortably over most prescription eyewear
  • Economically priced
Compliance
Meets ANSI Z87.1+ 2015 and CSA Z94.3 standards.
Specifications
Specifications
Anti-fog
Uvextra
Clear
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.