Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MARCH10 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$206.00 - $487.50
Specifications
| Antigen | MARCH10 |
|---|---|
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP19832720
![]() |
Novus Biologicals
NBP19832720UL |
20 μL |
Each for $206.00
|
|
|||||
NBP198327
![]() |
Novus Biologicals
NBP198327 |
100 μL |
Each for $487.50
|
|
|||||
Description
44265 Polyclonal specifically detects 44265 in Rat samples. It is validated for Western Blot.Specifications
| MARCH10 | |
| Polyclonal | |
| Rabbit | |
| NP_001013995 | |
| 162333 | |
| IgG | |
| 86 kDa |
| Western Blot | |
| Unconjugated | |
| RUO | |
| EC 6.3.2.-, FLJ35757, MARCH-Xprobable E3 ubiquitin-protein ligase MARCH10, membrane-associated ring finger (C3HC4) 10, Membrane-associated RING finger protein 10, Membrane-associated RING-CH protein X, RING finger protein 190RNF190 | |
| 43169 | |
| Primary |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title