Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

VEGF 165 Antibody (Bevacizumab) - Chimeric - Azide and BSA Free, Novus Biologicals™
SDP

Catalog No. NBP259638 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP259638 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP259638 Supplier Novus Biologicals Supplier No. NBP259638
Only null left
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

VEGF 165 Monoclonal specifically detects VEGF 165 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunoprecipitation, Block/Neutralize.

Specifications

Antigen VEGF 165
Applications Western Blot, Flow Cytometry, ELISA, Immunoprecipitation, Blocking Assay
Classification Monoclonal
Clone Bevacizumab
Conjugate Unconjugated
Dilution Western Blot, Flow Cytometry, ELISA, Immunoprecipitation, Blocking/Neutralizing
Formulation PBS with 0.02% Proclin 300. with 0.02% Proclin 300
Gene Alias biosimilar, biosimilars, MVCD1, vascular endothelial growth factor A, Vascular permeability factor, VEGF-A, VEGFMGC70609, VPFvascular endothelial growth factor
Gene Symbols VEGFA
Host Species Rabbit
Immunogen Recombinant human VEGF
Purification Method Protein A purified
Quantity 200 μg
Regulatory Status RUO
Research Discipline Angiogenesis, Biologically Active Proteins
Primary or Secondary Primary
Gene ID (Entrez) 7422
Test Specificity This antibody binds native and reduced human VEGF (isoform 165).
Target Species Human
Content And Storage Store at 4C for up to 3 months. For longer storage, aliquot and store at -20C.
Form Purified
Isotype IgG κ
Show More Show Less

For Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.