Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

VEGF-C Antibody (107/F10) - Azide and BSA Free, Novus Biologicals™
SDP

Catalog No. NB018377 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.02 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB018377 0.02 mg
NBP118626 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB018377 Supplier Novus Biologicals Supplier No. NBP1186260.02MG
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

VEGF-C Monoclonal antibody specifically detects VEGF-C in Human, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen).

Specifications

Antigen VEGF-C
Applications Immunohistochemistry, Western Blot, Immunocytochemistry, Immunohistochemistry (Frozen), Immunofluorescence
Classification Monoclonal
Clone 107/F10
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1-5 ug/ml, Immunohistochemistry 2-10 ug/ml, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunohistochemistry-Frozen 2-10 ug/ml
Formulation PBS (pH 7.2) with No Preservative
Gene Alias Flt4 ligand, Flt4-L, vascular endothelial growth factor C, Vascular endothelial growth factor-related protein, VEGF-C, VRPFLT4 ligand DHM
Gene Symbols VEGFC
Host Species Mouse
Immunogen Recombinant human VEGF-C
Purification Method Protein G purified
Quantity 0.02 mg
Regulatory Status RUO
Research Discipline Angiogenesis, Cancer
Primary or Secondary Primary
Gene ID (Entrez) 7424
Test Specificity This antibody detects human and rat VEGF-C in Western blots. No cross-reactivity is shown with VEGF-A.
Reconstitution Centrifuge vial prior to opening. Reconstitute in sterile water to a concentration of 0.1-1.0 mg/ml.
Target Species Human
Content And Storage Store at -80C in the dark. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2 α
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.