Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

VEGF-C Antibody, Novus Biologicals™
SDP

Catalog No. p-7199462 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB018001 0.025 mg
NB11061022 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB018001 Supplier Novus Biologicals Supplier No. NB110610220.025MG
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody has been used in 11 publications

VEGF-C Polyclonal specifically detects VEGF-C in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.

Specifications

Antigen VEGF-C
Applications Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot, Immunohistochemistry
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1:500-1:1000, Immunohistochemistry 1:50-200, Immunohistochemistry-Paraffin 1:50-1:200, Immunohistochemistry-Frozen 1:10-1:500
Formulation A 0.2 um filtered solution in PBS with No Preservative
Gene Accession No. O35757
Gene Alias Flt4 ligand, Flt4-L, vascular endothelial growth factor C, Vascular endothelial growth factor-related protein, VEGF-C, VRPFLT4 ligand DHM
Gene Symbols VEGFC
Host Species Rabbit
Immunogen This antibody was produced by immunizing rabbits with purified rat VEGF-C recombinant protein.
Purification Method Protein G purified
Quantity 0.025 mg
Regulatory Status RUO
Research Discipline Angiogenesis, Cancer
Primary or Secondary Primary
Gene ID (Entrez) 7424
Test Specificity This antibody recognizes rat VEGF-C and has strong cross-reactivity with mouse and human VEGF-C.
Reconstitution Reconstitute with 0.5 ml sterilized PBS to a final concentration of 0.2 mg/ml.
Target Species Rat, Human, Mouse
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.