Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
VG5Q Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 1 publication
$181.90 - $487.50
Specifications
Antigen | VG5Q |
---|---|
Dilution | Western Blot 1:500, ELISA reported in scientific literature, Immunohistochemistry 1:250, Immunocytochemistry/Immunofluorescence reported in scientific literature, Immunoprecipitation reported in scientific literature, Immunohistochemistry-Paraffin |
Applications | Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB100454SS
![]() |
Novus Biologicals
NB100454SS |
0.025 mL |
Each for $181.90
|
|
|||||
NB100454
![]() |
Novus Biologicals
NB100454 |
0.1 mL |
Each for $487.50
|
|
|||||
Description
VG5Q Polyclonal specifically detects VG5Q in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin.Specifications
VG5Q | |
Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation | |
Unconjugated | |
Rabbit | |
Angiogenesis, Hypoxia | |
Whole antisera with No Preservative | |
Angiogenic factor VG5Q, angiogenic factor with G patch and FHA domains 1, FLJ10283hVG5Q, GPATC7G patch domain-containing protein 7, GPATCH7HUS84971, HSU84971, VG5QVasculogenesis gene on 5q protein | |
AGGF1 | |
IgG | |
Unpurified | |
This is specific for human VG5Q. |
Western Blot 1:500, ELISA reported in scientific literature, Immunohistochemistry 1:250, Immunocytochemistry/Immunofluorescence reported in scientific literature, Immunoprecipitation reported in scientific literature, Immunohistochemistry-Paraffin | |
Polyclonal | |
Antisera | |
RUO | |
Human | |
Q8N302 | |
55109 | |
A synthetic peptide mapping to an epitope within residues 500-600 of the human VG5Q protein. [UniProt# Q8N302] | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
100 kDa |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title