Learn More
Thermo Scientific™ VitroEase™ Training Kit PROMO
Catalog No. PIA51363PM
Change view
Click to view available options
Quantity:
25 Preparations
Catalog No. Quantity
PIA51363PM 25 Preparations
1 options
This item is not returnable. View return policy
Catalog No. PIA51363PM Supplier Thermo Scientific™ Supplier No. A51363
This item is not returnable. View return policy
The Thermo Scientific™ VitroEase™ Cryo-EM Training Kit provides a complete solution that includes detailed instructions for the successful preparation of grids for cryo-electron microscopy single particle analysis.
  • The VitroEase™ Cryo-EM Training Kit has been designed to provide the necessary components to get started with cryo-EM analysis of protein samples
  • Sample preparation is a major bottleneck in cryo-electron microscopy (cryo-EM) single particle analysis (SPA)
  • Currently, sample optimization is often performed in a non-systematic way, extending optimization time over multiple days or even weeks
  • Kit includes a positive control protein, apoferritin, that has been designed specifically to meet the stringent needs for high protein purity, minimal protein aggregation, and intact protein structure
  • Commonly used accessories for grid preparation are also included such as tweezers for manipulation, grids, and grid boxes for storage
  • Detailed step-by-step instructions guide the user through the lengthy vitrification process
  • Together the control and components may be used as a comparison for success for new users or for quality control for expert users
  • Grids and accessories are provided by trusted sources
Specifications
Content And Storage Sufficient for 25 grid preparations. Contains VitroEase Apoferritin Standard (Cat. No. A51362), tweezers, grids, and grid boxes.
For Use With (Application) Cryo-EM SPA
For Use With (Equipment) Cryo-Electron Microscopes
Quantity 25 Preparations