Learn More
Restek™ Viva™ HPLC Columns with C18 Column Packing, 3μm Particle Size
Shop All Restek Corporation Products

Description
- Rugged, spherical particles with 300 angstrom pore size
- High proportion of pore/surface area available to large molecules
- General-purpose Restek C18 is a conventional monomeric octadecylsilane column
- Particle Size: 3μm
- USP Phase Code: L1
Specifications
Specifications
| Diameter (Metric) | 4.6 mm |
| Film Thickness | 3 μm |
| Flow Rate | 1.5 mL/min |
| For Use With | Proteins and Other Higher Molecular Weight Compounds |
| Length (Metric) | 30 mm |
| Particle Shape | Spherical |
| Particle Size | 3 μm |
| pH | 2.5 to 10 |
| Pore Size | 300 Å |
| Temperature | 80°C |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.