Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Restek™ Viva™ HPLC Columns with C18 Column Packing, 5μm Particle Size
SDP

Catalog No. 06739216 Shop All Restek Corporation Products
Encompass_Preferred
Change view
Click to view available options
Length (Metric):
30 mm
50 mm
100 mm
150 mm
200 mm
250 mm
Diameter (Metric):
1 mm
2.1 mm
4.6 mm
16 product options available for selection
Product selection table with 16 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (Metric) Diameter (Metric)
06-739-216 50 mm 1 mm
06-739-213 30 mm 2.1 mm
06-739-223 150 mm 4.6 mm
06-739-207 100 mm 4.6 mm
06-739-224 250 mm 1 mm
06-739-215 30 mm 4.6 mm
06-739-227 250 mm 4.6 mm
06-739-219 50 mm 4.6 mm
06-739-220 150 mm 1 mm
06-739-217 50 mm 2.1 mm
06-739-211 200 mm 4.6 mm
06-739-204 100 mm 1 mm
06-739-225 250 mm 2.1 mm
06-739-205 100 mm 2.1 mm
06-739-209 200 mm 2.1 mm
06-739-221 150 mm 2.1 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 16 options available.
16 options
Catalog No. 06-739-216 Supplier Restek™ Supplier No. 9514551
Add to Cart
Add to Cart

Wide bore silica columns excellent for general-purpose applications and separating peptides or proteins

  • Highly base-deactivated wide pore packing exhibits excellent peak shape
  • Rugged, spherical particles
  • 300Å pore size with a narrow pore size distribution—high proportion of pore/surface area available for large molecule and biomolecule separations
  • Carbon load: 9%
  • pH range: 2.5 to 10 pH range
  • Temperature limit: 80°C

Specifications

Diameter (Metric) 1 mm
Film Thickness 5 μm
Flow Rate 0.05 mL/min
For Use With Analyzing Large Molecules and Biomolecules
Length (Metric) 50 mm
Ligand Type Monomeric C18
Max. Pressure 4000 psi (275 bar)
Particle Shape Spherical
Particle Size 5 μm
pH 2.5 to 10
Pore Size 300 Å
Temperature 80°C
USP Type L1
Stationary Phase C18
Packing Material Silica
Carbon Load 9%
Column Type Reversed Phase
Endcapped Yes
Phase Reversed Phase
Material Stainless Steel
Product Type Column
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.