Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
VSIG3/IGSF11 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$206.00 - $487.50
Specifications
Antigen | VSIG3/IGSF11 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP15950320
![]() |
Novus Biologicals
NBP15950320UL |
20 μL |
Each for $206.00
|
|
|||||
NBP159503
![]() |
Novus Biologicals
NBP159503 |
100 μL |
Each for $487.50
|
|
|||||
Description
VSIG3/IGSF11 Polyclonal specifically detects VSIG3/IGSF11 in Human samples. It is validated for Western Blot.Specifications
VSIG3/IGSF11 | |
Polyclonal | |
Rabbit | |
Q5DX21 | |
152404 | |
Synthetic peptides corresponding to IGSF11(immunoglobulin superfamily, member 11) The peptide sequence was selected from the middle region of IGSF11. Peptide sequence EKLDNTLKLPPTATQDQVQGTVTIRNISALSSGLYQCVASNAIGTSTCLL. | |
Primary |
Western Blot | |
Unconjugated | |
RUO | |
Brain and testis-specific immunoglobulin superfamily protein, Bt-IGSF, BTIGSF, cancer/testis antigen 119, CT119, CXADR like 1, CXADRL1, IgSF11, Igsf13, immunoglobulin superfamily member 11, immunoglobulin superfamily, member 11, MGC35227, V-set and immunoglobulin domain containing 3, V-set and immunoglobulin domain-containing protein 3, VSIG3brain and testis-specific immunoglobin superfamily protein | |
IGSF11 | |
IgG |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title