Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tocris Bioscience™ VU 10010

Click to view available options
Quantity:
10 mg
50 mg
Description
Selective allosteric potentiator of M4 acetylcholine receptors (EC50 values are 33 and 0.7 nM for ACh in the absence and presence of VU10010 respectively). Binds to an allosteric site on the receptor increasing affinity for ACh and coupling to G-proteins.
Specifications
Specifications
| Quantity | 10 mg |
| Formula Weight | 345.85 |
| Purity | >99% |
| Molecular Weight (g/mol) | 345.845 |
| Chemical Name or Material | VU 10010 |
Safety and Handling
| Recommended Storage | Store at +4°C |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction