Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytiva Whatman™ Anotop™ IC 25mm Syringe Filter

Catalog No. 09302125 Shop All Cytiva Products
Click to view available options
:
Pore Size: 0.2 μm, Diameter: 10 mm, 250/Pk.

Can be used with most organic solvents and aqueous materials and are suitable for sample volumes up to 100mL

The distinctive hexagonal housing is manufactured from pigment-free polypropylene to eliminate sample contamination. No wetting agents or adhesives are used in the manufacturing process.
Anotop Syringe Filters contain the proprietary alumina-based Anopore membrane. Glass microfiber prefilter versions are available for difficult-to-filter samples.

  • Each batch certified for ion chromatography (IC)
  • Enhanced consistency of analytical results
  • Extended column life
  • Certified and guaranteed low levels of anion leaching for improved results
  • Inlet: female Luer-Lok™
  • Outlet: male slip Luer
  • Use for HPLC aqueous/organic sample preparation, cold sterilization of growth media, solvent filtration for spectroanalysis, phage and virus filtration, liposome extrusion, removal of high molecular weight proteins or polymers, colloid removal from deionized water

TRUSTED_SUSTAINABILITY

Specifications

Material Polypropylene
Pore Size 0.2 μm
Sterility Non-sterile
Quantity 250/Pk.
Type IC Syringe Filter
Diameter (Metric) 10 mm
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.