Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cytiva Whatman™ Mini-UniPrep™ Multicompressor and Accessories
Shop All Cytiva Products
Click to view available options
Type:
Multicompressor
Tray
Description
- Streamlines syringeless filtration by filtering up to eight filter vials simultaneously
- Designed for Cytiva's Mini-UniPrep™ Standard Plastic Filter Vial
- Mini-UniPrep™ Vials can be loaded directly into the autosampler
- The multiprocessor includes one blue 8-position tray
- Additional trays provide the added convenience of pre-loading up to eight Mini-UniPrep™ Filter Vials

Specifications
Specifications
Color | White, Purple |
For Use With (Application) | HPLC/UHPLC |
Includes | (1) 8-Position Tray for Standard Plastic Mini-UniPrep™ Filter Vials |
Product Line | Whatman™ |
Quantity | 1 Ea. |
Type | Multicompressor |
Dimensions (L x W x H) | 330 x 324 x 185 mm |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction