Learn More
Cytiva Whatman™ UniPrep™ Syringeless Filters - Glass Microfiber
Shop All Cytiva Products

Description
UniPrep syringeless filters are preassembled filtration devices for the filtration and storage of laboratory samples.
- Layered glass microfiber depth filter for use with samples containing aqueous or organic solvents (indicated pore size is the particle retention rating)
- Consists of external sample test tube, plunger (which incorporates membrane filter and prefilter), removable cap
- Filtration area: 0.3cm2
- Safe, efficient—prefilter and membrane can't pop off
- Air purge unnecessary—inserting plunger into tube provides a positive pressure purge of sample through filter
- Sealed glass microfiber prefilter traps larger particles before they can clog the membrane
- Increase lab productivity and efficiency
- Generate less waste
- Realize greater overall cost savings per test
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications
Specifications
Material (Membrane) | Glass Microfiber |
Pore Size | 0.45 μm |
Filtration Area | 0.28 cm2 |
Hold up Volume (After Air Purge) | 50 μL |
Type | Syringeless Filter |
Diameter (Metric) | 17 mm |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.