Learn More
Eisco™ White Quartzite Metamorphic Rock Specimen
SCI_ED
Catalog No. s27603 Shop All Eisco Products
Click to view available options
Quantity:
1/Ea.
12/Pk.
Quartzite is formed from sandstone that has been tightly compressed to the point where the clasts have recrystallized to form a new rock. This formation mechanism, coupled the inherent strength of quartz, makes quartzite a very durable rock.
  • Correlates well to Next Generation Science Standards Evidence
  • Useful in the classroom for lessons on metamorphic rocks
  • Ideal for identification & classification exercises
Specifications
Product Type Metamorphic Rock
Grade K to 16
Quantity 12/Pk.
Specimen Size 2 to 3 cm
Weight (Metric) 0.1 kg
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.