Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Wnt3 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$487.50
Specifications
Antigen | Wnt3 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP156996
![]() |
Novus Biologicals
NBP156996 |
100 μL |
Each for $487.50
|
|
|||||
NBP15699620
![]() |
Novus Biologicals
NBP15699620UL |
20 μL | N/A | N/A | N/A | ||||
Description
Wnt3 Polyclonal specifically detects Wnt3 in Human, Mouse samples. It is validated for Western Blot.Specifications
Wnt3 | |
Polyclonal | |
Rabbit | |
Stem Cell Signaling Pathway, Wnt Signaling Pathway | |
INT4Proto-oncogene Int-4 homolog, MGC131950, MGC138321, MGC138323, proto-oncogene Wnt-3, wingless-type MMTV integration site family, member 3, WNT-3 proto-oncogene protein | |
WNT3 | |
IgG | |
43 kDa |
Western Blot | |
Unconjugated | |
RUO | |
P56703 | |
7473 | |
Synthetic peptides corresponding to WNT3 (wingless-type MMTV integration site family, member 3) The peptide sequence was selected from the middle region of WNT3 (NP_110380). Peptide sequence SHHKGPPGEGWKWGGCSEDADFGVLVSREFADARENRPDARSAMNKHNNE. | |
Primary |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title