Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

wnvNS3 Protease Antibody [DyLight 650], Novus Biologicals™
SDP

Catalog No. AF2907W Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF2907W 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AF2907W Supplier Novus Biologicals Supplier No. AF2907W
Only null left
Add to Cart
Add to Cart

Goat Polyclonal Antibody

wnvNS3 Protease Polyclonal antibody specifically detects wnvNS3 Protease in Virus samples. It is validated for Immunoprecipitation,Western Blot

Specifications

Antigen wnvNS3 Protease
Applications Immunoprecipitation, Western Blot
Classification Polyclonal
Conjugate DyLight 650
Formulation 50mM Sodium Borate
Host Species Goat
Immunogen E. coli-derived recombinant wnvNS3 Protease, expressed as a fusion protein with cofactor NS2L, Gly1506-Leu1689, Accession # ABD85079
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 912267
Target Species Virus
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.