Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

xCT Antibody (23H6), Novus Biologicals™
SDP

Catalog No. p-200022427 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
50 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB298346 50 μL
NB298347 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB298346 Supplier Novus Biologicals Supplier No. NBP32631550UL
Only null left
Add to Cart
Add to Cart

Human Monoclonal Antibody

xCT Monoclonal antibody specifically detects xCT in Human samples. It is validated for ELISA

Specifications

Antigen xCT
Applications ELISA
Classification Monoclonal
Clone 23H6
Conjugate Unconjugated
Dilution ELISA
Formulation 50% Glycerol, 0.01M PBS, PH 7.4
Gene Alias Amino acid transport system xc-, Calcium channel blocker resistance protein CCBR1, CCBR1, cystine/glutamate transporter, SLC7A11, solute carrier family 7 (anionic amino acid transporter light chain, xc- system), member 11, Solute carrier family 7 member 11, solute carrier family 7, (cationic amino acid transporter, y+ system) member 11, xCT
Host Species Human
Immunogen Recombinant Human xCT protein [UniProt Q9UPY5]
Purification Method Affinity purified
Quantity 50 μL
Regulatory Status RUO
Research Discipline Amino Acids Drugs and other small molecules, Cancer, Cell Biology, Lipid and Metabolism, Neuronal Cell Markers, Neuroscience, Neurotransmission, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 23657
Target Species Human
Content And Storage Store at -20 to -70C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.